siRNA Details
| Gene Symbol: Trappc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177240.3 | NP_796214.2 | 20 | 2344 | ||
| GenBank mRNA | AK042816.1 | - | 3 | 755 | ||
| AK083445.1 | - | 20 | 2341 | |||
| AK160812.1 | - | 19 | 2668 | |||
| BC063065.1 | - | 13 | 1954 | |||
| BC094246.1 | - | 4 | 426 | |||
| BC147504.1 | - | 20 | 2262 | 9056419 | ||
| BC151159.1 | - | 20 | 2262 | 9088048 | ||

Trappc11
Genomic Map