siRNA Details
| Gene Symbol: St8sia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011374.2 | NP_035504.2 | 5 | 7434 | ||
| GenBank mRNA | AK039733.1 | - | 1 | 904 | ||
| AK040548.1 | - | 1 | 946 | |||
| AK079057.1 | - | 1 | 393 | |||
| BC110671.1 | - | 1 | 420 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St8sia1| Gene Symbol: St8sia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011374.2 | NP_035504.2 | 5 | 7434 | ||
| GenBank mRNA | AK039733.1 | - | 1 | 904 | ||
| AK040548.1 | - | 1 | 946 | |||
| AK079057.1 | - | 1 | 393 | |||
| BC110671.1 | - | 1 | 420 | |||