siRNA Details
| Gene Symbol: L3mbtl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177278.5 | NP_796252.2 | 4 | 671 | ||
| GenBank mRNA | AK039470.1 | - | 4 | 642 | ||
| AK053068.1 | - | 4 | 672 | |||
| BC147118.1 | - | 3 | 434 | |||
| BC147119.1 | - | 3 | 434 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

L3mbtl4| Gene Symbol: L3mbtl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177278.5 | NP_796252.2 | 4 | 671 | ||
| GenBank mRNA | AK039470.1 | - | 4 | 642 | ||
| AK053068.1 | - | 4 | 672 | |||
| BC147118.1 | - | 3 | 434 | |||
| BC147119.1 | - | 3 | 434 | |||