siRNA Details
| Gene Symbol: D830039M14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028369.1 | - | 1 | 1289 | ||
| AK039329.1 | - | 1 | 1034 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D830039M14Rik| Gene Symbol: D830039M14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028369.1 | - | 1 | 1289 | ||
| AK039329.1 | - | 1 | 1034 | |||