siRNA Details
| Gene Symbol: Snrnp200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177214.4 | NP_796188.2 | 2, 3 | 310 | ||
| GenBank mRNA | AK039567.1 | - | 2, 3 | 316 | ||
| AK081920.1 | - | 2, 3 | 329 | |||
| AK149291.1 | - | 2, 3 | 329 | |||
| BC063261.1 | - | 2, 3 | 310 | 6408607 | ||

Snrnp200
Genomic Map