siRNA Details
| Gene Symbol: CTSV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201575.1 | NP_001188504.1 | 3 | 260 | ||
| NM_001333.3 | NP_001324.2 | 3 | 463 | |||
| GenBank mRNA | AB001928.1 | - | 3 | 286 | ||
| AF070448.1 | - | 2 | 240 | |||
| AK310645.1 | - | 3 | 732 | |||
| AK310740.1 | - | 3 | 395 | |||
| AK312806.1 | - | 3 | 260 | |||
| AY358641.1 | - | 3 | 463 | |||
| BC023504.2 | - | 3 | 277 | 3139314 | ||
| BC110512.1 | - | 3 | 332 | 40031744 | ||
| Y14734.1 | - | 3 | 282 | |||

CTSV
Genomic Map