siRNA Details
| Gene Symbol: Atp2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177236.4 | NP_796210.2 | 3 | 698 | ||
| GenBank mRNA | AK032322.1 | - | 3 | 623 | ||
| AK045516.1 | - | 3 | 698 | |||
| AK047008.1 | - | 1 | 3177 | |||
| BC118975.1 | - | 2 | 430 | 40098391 | ||
| BC118976.1 | - | 2 | 430 | 40098398 | ||

Atp2b3
Genomic Map