siRNA Details
| Gene Symbol: Trappc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177240.3 | NP_796214.2 | 20, 21 | 2408 | ||
| GenBank mRNA | AK042816.1 | - | 3, 4 | 819 | ||
| AK083445.1 | - | 20, 21 | 2405 | |||
| AK160812.1 | - | 19, 20 | 2732 | |||
| BC063065.1 | - | 13, 14 | 2018 | |||
| BC094246.1 | - | 4, 5 | 490 | |||
| BC147504.1 | - | 20, 21 | 2326 | 9056419 | ||
| BC151159.1 | - | 20, 21 | 2326 | 9088048 | ||

Trappc11
Genomic Map