siRNA Details
| Gene Symbol: CTSO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001334.2 | NP_001325.1 | 4 | 534 | ||
| GenBank mRNA | AK300112.1 | - | 2 | 157 | ||
| AK308323.1 | - | 4 | 540 | |||
| AK312354.1 | - | 4 | 499 | |||
| BC049206.1 | - | 4 | 487 | 5189657 | ||
| BI834113.1 | - | 4 | 537 | |||
| X77383.1 | - | 4 | 534 | |||

CTSO
Genomic Map