siRNA Details
Gene Symbol: D830039M14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028369.1 | - | 1 | 1373 | ||
AK039329.1 | - | 1 | 1118 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D830039M14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028369.1 | - | 1 | 1373 | ||
AK039329.1 | - | 1 | 1118 |