siRNA Details
| Gene Symbol: Zfp629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177226.5 | NP_796200.1 | 3 | 5108 | ||
| GenBank mRNA | AK172933.1 | - | 4 | 4838 | ||
| BC035547.1 | - | 3 | 4750 | 4481313 | ||
| BC067053.1 | - | 3 | 5108 | 30543652 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp629| Gene Symbol: Zfp629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177226.5 | NP_796200.1 | 3 | 5108 | ||
| GenBank mRNA | AK172933.1 | - | 4 | 4838 | ||
| BC035547.1 | - | 3 | 4750 | 4481313 | ||
| BC067053.1 | - | 3 | 5108 | 30543652 | ||