siRNA Details
| Gene Symbol: Ccdc158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177230.3 | NP_796204.1 | 15, 16 | 2475 | ||
| GenBank mRNA | AK029997.1 | - | 15, 16 | 2475 | ||
| BC137976.1 | - | 15, 16 | 2444 | 8860995 | ||
| BC145072.1 | - | 15, 16 | 2445 | |||

Ccdc158
Genomic Map