siRNA Details
| Gene Symbol: Fam19a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177233.5 | NP_796207.2 | 6 | 1656 | ||
| GenBank mRNA | AK081531.1 | - | 6 | 1640 | ||
| AK149355.1 | - | 6 | 1659 | |||
| BC089490.1 | - | 6 | 1616 | 30504398 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam19a4| Gene Symbol: Fam19a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177233.5 | NP_796207.2 | 6 | 1656 | ||
| GenBank mRNA | AK081531.1 | - | 6 | 1640 | ||
| AK149355.1 | - | 6 | 1659 | |||
| BC089490.1 | - | 6 | 1616 | 30504398 | ||