siRNA Details
| Gene Symbol: Klhl31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172925.2 | NP_766513.2 | 3 | 5098 | ||
| GenBank mRNA | AK036669.1 | - | 1 | 1637 | ||
| AK085911.1 | - | 1 | 2302 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177238.1 | - | 2302 | Chr. - - [-] on Build | ||

Klhl31
Genomic Map