siRNA Details
| Gene Symbol: Pptc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177242.4 | NP_796216.2 | 6 | 1455 | ||
| GenBank mRNA | AK078914.1 | - | 6 | 1463 | ||
| AK083481.1 | - | 6 | 1472 | |||
| AK135118.1 | - | 6 | 1488 | |||
| AK162261.1 | - | 6 | 1020 | |||
| BC038622.1 | - | 6 | 1095 | |||
| BC068149.1 | - | 6 | 1455 | 30537082 | ||

Pptc7
Genomic Map