siRNA Details
Gene Symbol: 9230112E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033807.1 | - | 802 | |||
BC057100.1 | - | 726 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9230112E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033807.1 | - | 802 | |||
BC057100.1 | - | 726 |