siRNA Details
| Gene Symbol: Pisd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177298.3 | NP_796272.2 | 3, 4 | 333 | ||
| GenBank mRNA | AK033483.1 | - | 3, 4 | 325 | ||
| AK034656.1 | - | 3, 4 | 335 | |||
| AK162028.1 | - | 3, 4 | 326 | |||
| AK162737.1 | - | 3, 4 | 334 | |||
| AK167554.1 | - | 2, 3 | 275 | |||
| BC070408.1 | - | 3, 4 | 332 | 6809528 | ||
| BC094594.1 | - | 3, 4 | 324 | 5707749 | ||
| BC116695.1 | - | 3, 4 | 335 | 40105525 | ||
| BC117509.1 | - | 3, 4 | 335 | 40105529 | ||
| CX201846.1 | - | 3, 4 | 348 | |||

Pisd
Genomic Map