siRNA Details
| Gene Symbol: Plcxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177355.3 | NP_796329.2 | 2 | 599 | ||
| GenBank mRNA | AK038575.1 | - | 3 | 690 | ||
| AK044972.1 | - | 2 | 446 | |||
| BC137637.1 | - | 2 | 454 | 8860656 | ||
| BC137639.1 | - | 2 | 454 | 8860658 | ||

Plcxd3
Genomic Map