siRNA Details
| Gene Symbol: Tarm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177363.3 | NP_796337.1 | 3 | 281 | ||
| GenBank mRNA | AK036904.1 | - | 3 | 282 | ||
| AK089206.1 | - | 274 | ||||
| AK155804.1 | - | 3 | 289 | |||
| AK170277.1 | - | 2 | 212 | |||
| DQ973493.1 | - | 3 | 233 | |||

Tarm1
Genomic Map