siRNA Details
| Gene Symbol: Ofd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177429.3 | NP_803178.2 | 3 | 617 | ||
| GenBank mRNA | AJ438159.1 | - | 3 | 468 | ||
| BC119070.1 | - | 3 | 300 | 8733819 | ||
| BC120487.1 | - | 3 | 300 | 40129410 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ofd1| Gene Symbol: Ofd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177429.3 | NP_803178.2 | 3 | 617 | ||
| GenBank mRNA | AJ438159.1 | - | 3 | 468 | ||
| BC119070.1 | - | 3 | 300 | 8733819 | ||
| BC120487.1 | - | 3 | 300 | 40129410 | ||