siRNA Details
| Gene Symbol: DPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039589.1 | NP_001034678.1 | 3 | 506 | ||
| NM_001319165.1 | NP_001306094.1 | 3 | 496 | |||
| NM_001319166.1 | NP_001306095.1 | 3 | 506 | |||
| NM_001319167.1 | NP_001306096.1 | 3 | 563 | |||
| NM_001319168.1 | NP_001306097.1 | 3 | 454 | |||
| NM_001319170.1 | NP_001306099.1 | 3 | 566 | |||
| NM_001319171.1 | NP_001306100.1 | 3 | 506 | |||
| NM_001384.4 | NP_001375.2 | 3 | 506 | |||
| XM_017000502.1 | XP_016855991.1 | 3 | 508 | |||
| XM_017000503.1 | XP_016855992.1 | 3 | 442 | |||
| XM_017000504.1 | XP_016855993.1 | 3 | 550 | |||
| XM_017000505.1 | XP_016855994.1 | 3 | 482 | |||
| XM_017000506.1 | XP_016855995.1 | 3 | 550 | |||
| GenBank mRNA | AF053003.1 | - | 3 | 472 | ||
| AK091303.1 | - | 3 | 425 | |||
| AK315506.1 | - | 3 | 490 | |||
| BC001389.2 | - | 3 | 465 | 3051455 | ||
| BC003181.1 | - | 3 | 483 | 2958202 | ||
| BC016956.1 | - | 3 | 465 | 3900854 | ||
| BG385934.1 | - | 3 | 491 | |||
| BG472282.1 | - | 3 | 524 | |||
| BG747150.1 | - | 3 | 543 | |||
| BM564151.1 | - | 457 | ||||
| BM794530.1 | - | 3 | 448 | |||
| BQ056213.1 | - | 3 | 480 | |||
| BQ215136.1 | - | 3 | 514 | |||
| BT007431.1 | - | 3 | 311 | |||
| CX164803.1 | - | 3 | 433 | |||
| DA906783.1 | - | 3 | 508 | |||

DPH2
Genomic Map