siRNA Details
| Gene Symbol: A530053G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080052.1 | - | 2, 3 | 300 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177329.1 | - | 299 | Chr. - - [-] on Build | ||

A530053G22Rik
Genomic Map