siRNA Details
| Gene Symbol: Zfp945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110254.1 | NP_001103724.1 | 5 | 2574 | ||
| NM_177358.4 | NP_796332.2 | 6 | 2695 | |||
| GenBank mRNA | AK034838.1 | - | 1 | 1086 | ||
| BC078460.1 | - | 6 | 2665 | 6851563 | ||

Zfp945
Genomic Map