siRNA Details
| Gene Symbol: Scgb2b24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177446.2 | NP_803229.1 | 2 | 207 | ||
| GenBank mRNA | AA794526.1 | - | 2 | 207 | ||
| AY370635.1 | - | 2 | 224 | |||
| BC115602.1 | - | 2 | 207 | 40102624 | ||
| BC115603.1 | - | 2 | 207 | 40102625 | ||
| GU269236.1 | - | 2 | 197 | |||

Scgb2b24
Genomic Map