siRNA Details
| Gene Symbol: Rimkla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177572.4 | NP_808240.2 | 5 | 1109 | ||
| GenBank mRNA | AK047164.1 | - | 5 | 1104 | ||
| AK163360.1 | - | 5 | 1102 | |||
| BC057371.1 | - | 5 | 1090 | 6835897 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rimkla| Gene Symbol: Rimkla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177572.4 | NP_808240.2 | 5 | 1109 | ||
| GenBank mRNA | AK047164.1 | - | 5 | 1104 | ||
| AK163360.1 | - | 5 | 1102 | |||
| BC057371.1 | - | 5 | 1090 | 6835897 | ||