siRNA Details
| Gene Symbol: A530053G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079908.1 | - | 4 | 407 | ||
| AK080052.1 | - | 5 | 505 | |||
| BC051224.1 | - | 4 | 421 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177329.1 | - | 504 | Chr. - - [-] on Build | ||

A530053G22Rik
Genomic Map