siRNA Details
| Gene Symbol: Exoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177333.3 | NP_796307.2 | 9 | 1625 | ||
| GenBank mRNA | AK088333.1 | - | 9 | 1619 | ||
| AK089979.1 | - | 9 | 1635 | |||
| AK131141.1 | - | 9 | 1609 | |||
| AK141537.1 | - | 9 | 1625 | |||
| AK187380.1 | - | 2 | 97 | |||
| BC031743.1 | - | 9 | 1613 | 4187527 | ||

Exoc3
Genomic Map