siRNA Details
| Gene Symbol: 9230109A22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033789.1 | - | 3 | 1163 | ||
| AK039160.1 | - | 4 | 1075 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9230109A22Rik| Gene Symbol: 9230109A22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033789.1 | - | 3 | 1163 | ||
| AK039160.1 | - | 4 | 1075 | |||