siRNA Details
| Gene Symbol: Hykk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177351.4 | NP_796325.2 | 5 | 2324 | ||
| GenBank mRNA | AK085527.1 | - | 5 | 2334 | ||
| AK138851.1 | - | 5 | 2310 | |||
| AK162425.1 | - | 5 | 2325 | |||
| BC038618.1 | - | 5 | 2303 | 4221933 | ||

Hykk
Genomic Map