siRNA Details
| Gene Symbol: Plcxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038575.1 | - | 3 | 1860 | ||
| BC137637.1 | - | 2 | 1628 | 8860656 | ||
| BC137639.1 | - | 2 | 1626 | 8860658 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plcxd3| Gene Symbol: Plcxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038575.1 | - | 3 | 1860 | ||
| BC137637.1 | - | 2 | 1628 | 8860656 | ||
| BC137639.1 | - | 2 | 1626 | 8860658 | ||