siRNA Details
| Gene Symbol: Zfp799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177359.5 | NP_796333.3 | 4 | 3798 | ||
| GenBank mRNA | AK031686.1 | - | 4 | 3786 | ||
| AK083640.1 | - | 4 | 3775 | |||
| AK137037.1 | - | 5 | 3824 | |||
| AK148571.1 | - | 1 | 1690 | |||
| BC096552.1 | - | 4 | 3748 | 30618827 | ||

Zfp799
Genomic Map