siRNA Details
| Gene Symbol: Slfnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177570.3 | NP_808238.1 | 4 | 1434 | ||
| GenBank mRNA | AK077106.1 | - | 4 | 1435 | ||
| BC117545.1 | - | 3 | 1182 | 40105640 | ||
| BC118052.1 | - | 3 | 1185 | 40105639 | ||

Slfnl1
Genomic Map