siRNA Details
| Gene Symbol: 9930104L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037053.1 | - | 3 | 1039 | ||
| BC117984.1 | - | 1 | 2058 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9930104L06Rik| Gene Symbol: 9930104L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037053.1 | - | 3 | 1039 | ||
| BC117984.1 | - | 1 | 2058 | |||