siRNA Details
| Gene Symbol: Dcdc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195617.1 | NP_001182546.1 | 1 | 395 | ||
| NM_177577.3 | NP_808245.2 | 1 | 395 | |||
| GenBank mRNA | AK085257.1 | - | 1 | 209 | ||
| AK220370.1 | - | 1 | 168 | |||
| BC045136.1 | - | 1 | 395 | 6306251 | ||

Dcdc2a
Genomic Map