siRNA Details
| Gene Symbol: March11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177597.5 | NP_808265.2 | 1, 2 | 729 | ||
| GenBank mRNA | AK035990.1 | - | 1, 2 | 754 | ||
| AK162044.1 | - | 1, 2 | 724 | |||
| BC139422.1 | - | 1, 2 | 681 | 8862444 | ||
| BC139423.1 | - | 1, 2 | 681 | 8862445 | ||

March11
Genomic Map