siRNA Details
| Gene Symbol: Gm4814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050630.1 | - | 3 | 588 | ||
| BC134387.1 | - | 3 | 479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4814| Gene Symbol: Gm4814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050630.1 | - | 3 | 588 | ||
| BC134387.1 | - | 3 | 479 | |||