siRNA Details
| Gene Symbol: Igfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177642.3 | NP_808310.2 | 25 | 7928 | ||
| GenBank mRNA | AK036509.1 | - | 6 | 703 | ||
| BC107027.1 | - | 3 | 252 | 40050318 | ||
| BC107028.2 | - | 3 | 252 | 40050319 | ||
| EU429481.1 | - | 11 | 4800 | |||
| EU429482.1 | - | 10 | 1667 | |||
| EU429483.2 | - | 20 | 3081 | |||
| GU290042.1 | - | 25 | 7928 | |||

Igfn1
Genomic Map