siRNA Details
| Gene Symbol: Ttc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177667.4 | NP_808335.1 | 2 | 697 | ||
| GenBank mRNA | AK028895.1 | - | 2 | 698 | ||
| BC096484.1 | - | 2 | 676 | |||
| BC132162.1 | - | 2 | 638 | 40130439 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc22| Gene Symbol: Ttc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177667.4 | NP_808335.1 | 2 | 697 | ||
| GenBank mRNA | AK028895.1 | - | 2 | 698 | ||
| BC096484.1 | - | 2 | 676 | |||
| BC132162.1 | - | 2 | 638 | 40130439 | ||