siRNA Details
| Gene Symbol: Zfp637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177684.2 | NP_808352.1 | 3 | 220 | ||
| GenBank mRNA | AB221671.1 | - | 2 | 123 | ||
| AK146789.1 | - | 3 | 353 | |||
| BC021601.1 | - | 3 | 249 | 5150384 | ||
| BC037994.1 | - | 2 | 203 | |||
| BC048843.1 | - | 3 | 220 | 5031082 | ||

Zfp637
Genomic Map