siRNA Details
Gene Symbol: Gm4876 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047082.1 | - | 1 | 145 | ||
BC120768.1 | - | 1 | 121 | 40129691 | ||
BC120770.1 | - | 1 | 124 | 40129693 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4876 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047082.1 | - | 1 | 145 | ||
BC120768.1 | - | 1 | 121 | 40129691 | ||
BC120770.1 | - | 1 | 124 | 40129693 |