siRNA Details
| Gene Symbol: Ppm1n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177691.3 | NP_808359.1 | 1 | 801 | ||
| GenBank mRNA | AK044232.1 | - | 1 | 801 | ||
| AK044334.1 | - | 1 | 802 | |||
| BC096372.1 | - | 1 | 790 | 4507828 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppm1n| Gene Symbol: Ppm1n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177691.3 | NP_808359.1 | 1 | 801 | ||
| GenBank mRNA | AK044232.1 | - | 1 | 801 | ||
| AK044334.1 | - | 1 | 802 | |||
| BC096372.1 | - | 1 | 790 | 4507828 | ||