siRNA Details
Gene Symbol: Gm9871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK027905.1 | - | 4, 5 | 962 | ||
BC125571.1 | - | 3, 4 | 595 | |||
BC144923.1 | - | 3, 4 | 713 | |||
BC145947.1 | - | 3, 4 | 595 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm9871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK027905.1 | - | 4, 5 | 962 | ||
BC125571.1 | - | 3, 4 | 595 | |||
BC144923.1 | - | 3, 4 | 713 | |||
BC145947.1 | - | 3, 4 | 595 |