siRNA Details
| Gene Symbol: AA986860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177604.3 | NP_808272.2 | 4 | 1612 | ||
| GenBank mRNA | AK028957.1 | - | 4 | 1503 | ||
| AK053179.1 | - | 4 | 1614 | |||
| AK085225.1 | - | 2 | 1909 | |||
| AK086121.1 | - | 4 | 1487 | |||
| BC099401.1 | - | 1 | 551 | 30713499 | ||
| BC117765.1 | - | 1 | 825 | 40090933 | ||
| BC117766.1 | - | 1 | 825 | 40090934 | ||
| BC129959.1 | - | 1 | 824 | |||
| BC129960.1 | - | 1 | 825 | 40090940 | ||

AA986860
Genomic Map