siRNA Details
| Gene Symbol: Pdzd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195265.1 | NP_001182194.1 | 16 | 3143 | ||
| GenBank mRNA | AK033653.1 | - | 6 | 1038 | ||
| BC096488.1 | - | 6 | 1259 | |||
| BC147398.1 | - | 3 | 946 | 9056452 | ||
| BC147399.1 | - | 3 | 946 | 9056443 | ||
| BC172054.1 | - | 3 | 946 | |||
| KF041446.1 | - | 17 | 3109 | |||
| KF041447.1 | - | 17 | 3138 | |||
| KF041448.1 | - | 14 | 2923 | |||
| KF041449.1 | - | 15 | 2956 | |||
| KF041450.1 | - | 16 | 3048 | |||
| KF041451.1 | - | 17 | 2929 | |||

Pdzd7
Genomic Map