siRNA Details
| Gene Symbol: Plekhh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177606.4 | NP_808274.2 | 13 | 2267 | ||
| GenBank mRNA | AK029252.1 | - | 13 | 2269 | ||
| AK173337.1 | - | 10 | 1922 | |||
| BC125583.1 | - | 13 | 2232 | 40130098 | ||
| BC137804.1 | - | 13 | 2229 | 8860823 | ||

Plekhh2
Genomic Map