siRNA Details
| Gene Symbol: Rin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161365.1 | NP_001154837.1 | 2 | 184 | ||
| NM_177620.4 | NP_808288.2 | 2 | 295 | |||
| GenBank mRNA | AK032205.1 | - | 2 | 296 | ||
| AK133988.1 | - | 2 | 203 | |||
| AK154506.1 | - | 2 | 322 | |||
| AK154522.1 | - | 2 | 99 | |||
| AK154963.1 | - | 2 | 185 | |||
| AK166196.1 | - | 2 | 141 | |||
| AK167187.1 | - | 2 | 202 | |||
| AK171301.1 | - | 2 | 108 | |||
| AK171311.1 | - | 2 | 99 | |||
| BC137907.1 | - | 2 | 223 | 8860926 | ||

Rin3
Genomic Map