siRNA Details
| Gene Symbol: 4933409G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177651.3 | NP_808319.1 | 7, 8 | 464 | ||
| GenBank mRNA | AK077128.1 | - | 7, 8 | 465 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933409G03Rik| Gene Symbol: 4933409G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177651.3 | NP_808319.1 | 7, 8 | 464 | ||
| GenBank mRNA | AK077128.1 | - | 7, 8 | 465 | ||