siRNA Details
| Gene Symbol: 6820408C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289738.1 | NP_001276667.1 | 5, 6 | 711 | ||
| NM_177656.4 | NP_808324.1 | 5, 6 | 708 | |||
| GenBank mRNA | AK078482.1 | - | 5, 6 | 708 | ||
| BC137826.1 | - | 4, 5 | 588 | 8860845 | ||
| BC144943.1 | - | 4, 5 | 585 | |||
| BC144944.1 | - | 4, 5 | 588 | 9053492 | ||
| BC144945.1 | - | 4, 5 | 581 | |||

6820408C15Rik
Genomic Map