siRNA Details
| Gene Symbol: C130079G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177661.3 | NP_808329.1 | 6 | 1099 | ||
| GenBank mRNA | AK081819.1 | - | 6 | 1100 | ||
| Gene Symbol: Gm9696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140981.1 | - | 5 | 1475 | ||

C130079G13Rik
Genomic Map