siRNA Details
Gene Symbol: Gm4861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177665.3 | NP_808333.1 | 2 | 249 | ||
GenBank mRNA | AK079008.1 | - | 2 | 250 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177665.3 | NP_808333.1 | 2 | 249 | ||
GenBank mRNA | AK079008.1 | - | 2 | 250 |